Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 5G7]

Supplier: Abnova
H00009852-M02
10585-156EA 335 USD
10585-156
Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: 5G7]
Antibodies
Type: Primary
Antigen: EPM2AIP1
Clonality: Monoclonal
Clone: 5G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR