Anti-ZEB2 Mouse Monoclonal Antibody [clone: 2H5]

Supplier: Abnova
H00009839-M02
10606-094EA 335 USD
10606-094
Anti-ZEB2 Mouse Monoclonal Antibody [clone: 2H5]
Antibodies
Type: Primary
Antigen: ZEB2
Clonality: Monoclonal
Clone: 2H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR