Anti-GINS1 Mouse Polyclonal Antibody

Supplier: Abnova
H00009837-B02P
10557-484EA 335 USD
10557-484
Anti-GINS1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RP4-691N24.2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR