Anti-TRIM14 Rabbit Polyclonal Antibody

Supplier: Abnova
H00009830-D01P
10604-610EA 376 USD
10604-610
Anti-TRIM14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TRIM14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR