Anti-SPATA2 Mouse Monoclonal Antibody [clone: 1F1]

Supplier: Abnova
H00009825-M01
10601-930EA 335 USD
10601-930
Anti-SPATA2 Mouse Monoclonal Antibody [clone: 1F1]
Antibodies
Type: Primary
Antigen: SPATA2
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR