Anti-GIT2 Mouse Polyclonal Antibody

Supplier: Abnova
H00009815-A02
10557-494EA 243 USD
10557-494
Anti-GIT2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GIT2 (G protein-coupled receptor kinase interacting ArfGAP 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR