Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TOMM20 (translocase of outer mitochondrial membrane 20 homolog (yeast)) Mouse Polyclonal Antibody

Supplier: Abnova

H00009804-A01
10572-742EA 209.23 USD InStock
10572-742
Anti-TOMM20 (translocase of outer mitochondrial membrane 20 homolog (yeast)) Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TOMM20 (translocase of outer mitochondrial membrane 20 homolog (yeast))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty