Anti-KIAA0101 Mouse Monoclonal Antibody [clone: 3C11-1F11]

Supplier: Abnova
H00009768-M01
10591-020EA 335 USD
10591-020
Anti-KIAA0101 Mouse Monoclonal Antibody [clone: 3C11-1F11]
Antibodies
Type: Primary
Antigen: KIAA0101
Clonality: Monoclonal
Clone: 3C11-1F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR