Anti-HDAC4 Mouse Monoclonal Antibody [clone: 2B11]

Supplier: Abnova
H00009759-M03
10588-512EA 335 USD
10588-512
Anti-HDAC4 Mouse Monoclonal Antibody [clone: 2B11]
Antibodies
Type: Primary
Antigen: HDAC4
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR