Anti-KMT2B Mouse Monoclonal Antibody [clone: 4C10]

Supplier: Abnova
H00009757-M02
10593-544EA 335 USD
10593-544
Anti-KMT2B Mouse Monoclonal Antibody [clone: 4C10]
Antibodies
Type: Primary
Antigen: KMT2B
Clonality: Monoclonal
Clone: 4C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR