Anti-FAM65B Mouse Monoclonal Antibody [clone: 2F6-1A11]

Supplier: Abnova
H00009750-M02
10581-264EA 335 USD
10581-264
Anti-FAM65B Mouse Monoclonal Antibody [clone: 2F6-1A11]
Antibodies
Type: Primary
Antigen: FAM65B (family with sequence similarity 65, member B)
Clonality: Monoclonal
Clone: 2F6-1A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR