Anti-SLK Mouse Monoclonal Antibody [clone: 3E9]

Supplier: Abnova
H00009748-M01
10601-710EA 335 USD
10601-710
Anti-SLK Mouse Monoclonal Antibody [clone: 3E9]
Antibodies
Type: Primary
Antigen: SLK
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR