Anti-HERPUD1 Mouse Monoclonal Antibody [clone: 3E10]

Supplier: Abnova
H00009709-M01
10588-598EA 335 USD
10588-598
Anti-HERPUD1 Mouse Monoclonal Antibody [clone: 3E10]
Antibodies
Type: Primary
Antigen: HERPUD1
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR