Anti-ULK2 Mouse Monoclonal Antibody [clone: 50000]

Supplier: Abnova
H00009706-M10
10605-386EA 335 USD
10605-386
Anti-ULK2 Mouse Monoclonal Antibody [clone: 50000]
Antibodies
Type: Primary
Antigen: ULK2
Clonality: Monoclonal
Clone: 50000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR