Anti-CEP57 Rabbit Polyclonal Antibody

Supplier: Abnova
H00009702-D01P
10581-984EA 376 USD
10581-984
Anti-CEP57 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CEP57
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR