Anti-RAPGEF2 Mouse Monoclonal Antibody [clone: 1E8]

Supplier: Abnova
H00009693-M01
10599-160EA 335 USD
10599-160
Anti-RAPGEF2 Mouse Monoclonal Antibody [clone: 1E8]
Antibodies
Type: Primary
Antigen: RAPGEF2
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR