Anti-UBE3C Mouse Polyclonal Antibody

Supplier: Abnova
H00009690-A01
10573-354EA 243 USD
10573-354
Anti-UBE3C Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: UBE3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR