Anti-SAFB2 Mouse Monoclonal Antibody [clone: 4H3]

Supplier: Abnova
H00009667-M01
10599-284EA 335 USD
10599-284
Anti-SAFB2 Mouse Monoclonal Antibody [clone: 4H3]
Antibodies
Type: Primary
Antigen: SAFB2
Clonality: Monoclonal
Clone: 4H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR