Anti-PDE4DIP Mouse Polyclonal Antibody

Supplier: Abnova
H00009659-B01
10567-006EA 335 USD
10567-006
Anti-PDE4DIP Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDE4DIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR