Anti-IKBKE Mouse Monoclonal Antibody [clone: 1F7]

Supplier: Abnova
H00009641-M04
10590-078EA 335 USD
10590-078
Anti-IKBKE Mouse Monoclonal Antibody [clone: 1F7]
Antibodies
Type: Primary
Antigen: IKBKE (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon)
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR