Anti-IKBKE Mouse Monoclonal Antibody [clone: 2F1]

Supplier: Abnova
H00009641-M02
10590-074EA 335 USD
10590-074
Anti-IKBKE Mouse Monoclonal Antibody [clone: 2F1]
Antibodies
Type: Primary
Antigen: IKBKE (inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon)
Clonality: Monoclonal
Clone: 2F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR