Anti-ARHGEF10 Rabbit Polyclonal Antibody

Supplier: Abnova
H00009639-D01P
10577-238EA 376 USD
10577-238
Anti-ARHGEF10 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ARHGEF10 (Rho guanine nucleotide exchange factor (GEF) 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR