Anti-SNCAIP Mouse Monoclonal Antibody [clone: 2A8]

Supplier: Abnova
H00009627-M01
10591-504EA 335 USD
10591-504
Anti-SNCAIP Mouse Monoclonal Antibody [clone: 2A8]
Antibodies
Type: Primary
Antigen: SNCAIP
Clonality: Monoclonal
Clone: 2A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR