Anti-GUCA1C Rabbit Polyclonal Antibody

Supplier: Abnova
H00009626-D01
10588-164EA 376 USD
10588-164
Anti-GUCA1C Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GUCA1C (guanylate cyclase activator 1C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR