Anti-TRAF4 Mouse Monoclonal Antibody [clone: 3F6]

Supplier: Abnova
H00009618-M01
10604-562EA 335 USD
10604-562
Anti-TRAF4 Mouse Monoclonal Antibody [clone: 3F6]
Antibodies
Type: Primary
Antigen: TRAF4
Clonality: Monoclonal
Clone: 3F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR