Anti-RAB36 Mouse Monoclonal Antibody [clone: 6A6]

Supplier: Abnova
H00009609-M01
10569-206EA 335 USD
10569-206
Anti-RAB36 Mouse Monoclonal Antibody [clone: 6A6]
Antibodies
Type: Primary
Antigen: RAB36
Clonality: Monoclonal
Clone: 6A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR