Anti-IER2 Mouse Monoclonal Antibody [clone: 300]

Supplier: Abnova
H00009592-M05
10589-804EA 335 USD
10589-804
Anti-IER2 Mouse Monoclonal Antibody [clone: 300]
Antibodies
Type: Primary
Antigen: IER2
Clonality: Monoclonal
Clone: 300
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR