Anti-WTAP Mouse Monoclonal Antibody [clone: 1B11]

Supplier: Abnova
H00009589-M01
10605-844EA 335 USD
10605-844
Anti-WTAP Mouse Monoclonal Antibody [clone: 1B11]
Antibodies
Type: Primary
Antigen: WTAP
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR