Anti-MAD2L1BP Rabbit Polyclonal Antibody

Supplier: Abnova
H00009587-D01P
10592-160EA 376 USD
10592-160
Anti-MAD2L1BP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAD2L1BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR