Anti-ENTPD4 Mouse Monoclonal Antibody [clone: 4H7]

Supplier: Abnova
H00009583-M01
10585-044EA 335 USD
10585-044
Anti-ENTPD4 Mouse Monoclonal Antibody [clone: 4H7]
Antibodies
Type: Primary
Antigen: ENTPD4
Clonality: Monoclonal
Clone: 4H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR