Anti-CDC42BPB Mouse Monoclonal Antibody [clone: 2F4]

Supplier: Abnova
H00009578-M03
10581-648EA 335 USD
10581-648
Anti-CDC42BPB Mouse Monoclonal Antibody [clone: 2F4]
Antibodies
Type: Primary
Antigen: CDC42BPB
Clonality: Monoclonal
Clone: 2F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR