Anti-SPAG6 Mouse Monoclonal Antibody [clone: 2F3]

Supplier: Abnova
H00009576-M05
10601-918EA 335 USD
10601-918
Anti-SPAG6 Mouse Monoclonal Antibody [clone: 2F3]
Antibodies
Type: Primary
Antigen: SPAG6
Clonality: Monoclonal
Clone: 2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR