Anti-GDF3 Mouse Monoclonal Antibody [clone: 2B10]

Supplier: Abnova
H00009573-M01
10587-260EA 335 USD
10587-260
Anti-GDF3 Mouse Monoclonal Antibody [clone: 2B10]
Antibodies
Type: Primary
Antigen: GDF3 (growth differentiation factor 3)
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR