Anti-HIP1R Mouse Polyclonal Antibody

Supplier: Abnova
H00009026-B01P
10558-374EA 335 USD
10558-374
Anti-HIP1R Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HIP1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR