Anti-CLIC3 Mouse Monoclonal Antibody [clone: 2C5]

Supplier: Abnova
H00009022-M04
10582-388EA 335 USD
10582-388
Anti-CLIC3 Mouse Monoclonal Antibody [clone: 2C5]
Antibodies
Type: Primary
Antigen: CLIC3
Clonality: Monoclonal
Clone: 2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR