Anti-CLIC3 Rabbit Polyclonal Antibody

Supplier: Abnova
H00009022-D01P
10582-392EA 376 USD
10582-392
Anti-CLIC3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CLIC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR