Anti-SOCS3 Mouse Monoclonal Antibody [clone: 1F6]

Supplier: Abnova
H00009021-M03
10591-586EA 335 USD
10591-586
Anti-SOCS3 Mouse Monoclonal Antibody [clone: 1F6]
Antibodies
Type: Primary
Antigen: SOCS3
Clonality: Monoclonal
Clone: 1F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR