Anti-TAF1A Mouse Monoclonal Antibody [clone: 1F8]

Supplier: Abnova
H00009015-M01
10603-190EA 335 USD
10603-190
Anti-TAF1A Mouse Monoclonal Antibody [clone: 1F8]
Antibodies
Type: Primary
Antigen: TAF1A
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR