Anti-CDC42BPA Mouse Monoclonal Antibody [clone: 1G7]

Supplier: Abnova
H00008476-M01
10581-640EA 335 USD
10581-640
Anti-CDC42BPA Mouse Monoclonal Antibody [clone: 1G7]
Antibodies
Type: Primary
Antigen: CDC42BPA
Clonality: Monoclonal
Clone: 1G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR