Anti-RAD54L Mouse Monoclonal Antibody [clone: 4G2]

Supplier: Abnova
H00008438-M01
10599-080EA 335 USD
10599-080
Anti-RAD54L Mouse Monoclonal Antibody [clone: 4G2]
Antibodies
Type: Primary
Antigen: RAD54L
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR