Anti-HIST1H4H Mouse Monoclonal Antibody [clone: 6D4]

Supplier: Abnova
H00008365-M01
10588-864EA 335 USD
10588-864
Anti-HIST1H4H Mouse Monoclonal Antibody [clone: 6D4]
Antibodies
Type: Primary
Antigen: HIST1H4H
Clonality: Monoclonal
Clone: 6D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR