Anti-TCF7L2 Mouse Monoclonal Antibody [clone: 3D7]

Supplier: Abnova
H00006934-M03
10603-156EA 335 USD
10603-156
Anti-TCF7L2 Mouse Monoclonal Antibody [clone: 3D7]
Antibodies
Type: Primary
Antigen: TCF7L2
Clonality: Monoclonal
Clone: 3D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR