Anti-PRKACA Mouse Monoclonal Antibody [clone: 1D7]

Supplier: Abnova
H00005566-M02
10598-054EA 335 USD
10598-054
Anti-PRKACA Mouse Monoclonal Antibody [clone: 1D7]
Antibodies
Type: Primary
Antigen: PRKACA
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR