Anti-NEK2 Mouse Monoclonal Antibody [clone: 2F6]

Supplier: Abnova
H00004751-M01
10594-654EA 335 USD
10594-654
Anti-NEK2 Mouse Monoclonal Antibody [clone: 2F6]
Antibodies
Type: Primary
Antigen: NEK2 (NIMA-related kinase 2)
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR