Anti-SEPT2 Mouse Monoclonal Antibody [clone: 2F8]

Supplier: Abnova
H00004735-M06
10594-622EA 335 USD
10594-622
Anti-SEPT2 Mouse Monoclonal Antibody [clone: 2F8]
Antibodies
Type: Primary
Antigen: SEPT2
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR