Anti-NDUFV3 Rabbit Polyclonal Antibody

Supplier: Abnova
H00004731-D01P
10594-612EA 376 USD
10594-612
Anti-NDUFV3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFV3 (NADH dehydrogenase (ubiquinone) flavoprotein 3, 10kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR