Anti-NDUFS8 Mouse Polyclonal Antibody

Supplier: Abnova
H00004728-A01
10565-470EA 243 USD
10565-470
Anti-NDUFS8 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFS8 (NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR