Anti-NDUFS3 Mouse Monoclonal Antibody [clone: 1D6]

Supplier: Abnova
H00004722-M02
10593-486EA 335 USD
10593-486
Anti-NDUFS3 Mouse Monoclonal Antibody [clone: 1D6]
Antibodies
Type: Primary
Antigen: NDUFS3 (NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase))
Clonality: Monoclonal
Clone: 1D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR