Anti-NDUFS3 Rabbit Polyclonal Antibody

Supplier: Abnova
H00004722-D01P
10593-490EA 376 USD
10593-490
Anti-NDUFS3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFS3 (NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR