Anti-NDUFS3 Mouse Polyclonal Antibody

H00004722-B01
10565-462EA 335 USD
10565-462
Anti-NDUFS3 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NDUFS3 (NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR